cis-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(OC-6-22)-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate
AC36750-0010
AC36750-0050
AC36750-2500
AC36750-5000
TS36750-0010EA
139
USD
TS36750-0010
TS36750-0050
TS36750-2500
TS36750-5000
cis-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate 97%
cis-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate
Formula:
C₂₀H₁₈Cl₂N₄ORu MW: 502.361 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00150005 CAS Number: 98014-14-3 |
Specification Test Results
Appearance | Dark green to black Powder and/or chunks |
Infrared spectrum | Conforms |
Assay | ≥96.0 % (Based on Carbon) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...