3-[2-(2-Aminoethylamino)ethylamino]propyltrimethoxysilane tech.

Supplier: Thermo Scientific Chemicals
Danger

AC34617-0250 AC34617-1000 AC34617-5000
TS34617-0250EA 38 USD
TS34617-0250 TS34617-1000 TS34617-5000
3-[2-(2-Aminoethylamino)ethylamino]propyltrimethoxysilane tech.
3-[2-(2-Aminoethylamino)ethylamino]propyltrimethoxysilane
Formula: C₁₀H₂₇N₃O₃Si
Boiling Pt: 114…118 °C (2 mmHg)
Density: 1.031 g/cm³
Flash Pt: 137 °C
Storage Temperature: Ambient
MDL Number: MFCD00069173
CAS Number: 35141-30-1
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid (May discolor to pink to red to brown on storage)
Infrared spectrum Conforms
Refractive index 1.4570 to 1.4650 (20°C, 589 nm)
Titration with HClO4 14 to 18 % (as NH2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR