2-(4-Chlorosulfonylphenyl)ethyltrimethoxysilane 50% in dichloromethane

Supplier: Thermo Scientific Chemicals
Danger

AC33747-0050 AC33747-0250
TS33747-0050EA 124 USD
TS33747-0050 TS33747-0250
2-(4-Chlorosulfonylphenyl)ethyltrimethoxysilane 50% in dichloromethane
2-(4-Chlorosulfonylphenyl)ethyltrimethoxysilane
Formula: C₁₁H₁₇ClO₅SSi
Boiling Pt: >40 °C (760 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00054774
CAS Number: 126519-89-9
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear yellow to light brown Solution
Infrared spectrum Conforms
Assay 45 to 55 % solids

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR