Lithium isopropoxide 3 M in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC39650-1000 AC39650-8000
TS39650-1000EA 124 USD
TS39650-1000 TS39650-8000
Lithium isopropoxide 3 M in THF, AcroSeal™
Lithium isopropoxide
Formula: C₃H₇LiO
MW: 66.028 g/mol
Density: 0.87 g/cm³
Flash Pt: –19 °C
Storage Temperature: Ambient
MDL Number: MFCD00048286
CAS Number: 2388-10-5
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear yellow to red Solution
Strength 2.9 to 3.3 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR