Bis(2-methylallyl)-1,5-cyclooctadieneruthenium(II) (30 - 32% Ru)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bis-(2-methylallyl)cycloocta-1,5-diene ruthenium (II) complex, Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II), Bis(2-methylallyl)-1,5-cyclooctadieneruthenium (II)

AC29578-0010 AC29578-2500
TS29578-0010EA 165 USD
TS29578-0010 TS29578-2500
Bis(2-methylallyl)-1,5-cyclooctadieneruthenium(II) (30 - 32% Ru)
Bis(2-methylallyl)-1,5-cyclooctadieneruthenium(II)
Formula: C₁₆H₂₆Ru
Melting Pt: 81…85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00216965
CAS Number: 12289-94-0

Order Now

Specification Test Results

Appearance Beige to brown Crystalline powder
Infrared spectrum Conforms
Melting point 80 to 84 °C
NMR ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR