Cyclopentylmagnesium chloride 2 M in diethyl ether, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC20846-1000 AC20846-8000
TS20846-1000EA 79 USD
TS20846-1000 TS20846-8000
Cyclopentylmagnesium chloride 2 M in diethyl ether, AcroSeal™
Cyclopentylmagnesium chloride
Formula: C₅H₉ClMg
MW: 128.882 g/mol
Density: 0.86 g/cm³
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00001357
CAS Number: 32916-51-1
UN: 3399
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Clear to hazy yellow to gray to brown Solution (May develop some turbidity or precipitate)
Titration Grignard 1.9 to 2.2 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR