4-O-Methyldopamine hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC18774-0500
AC18774-5000
TS18774-0500EA
60
USD
TS18774-0500
TS18774-5000
4-O-Methyldopamine hydrochloride 98%
4-O-Methyldopamine hydrochloride
Formula:
C₉H₁₄ClNO₂ Melting Pt: 207…211 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012896 CAS Number: 645-33-0 |
Specification Test Results
Appearance | White to Light beige Powder |
Infrared spectrum | Conforms |
Melting point | 207 to 211 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...