Tributyltin trifluoromethanesulfonate 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tributyltin triflate

AC38893-0010 AC38893-0050
TS38893-0010EA 38 USD
TS38893-0010 TS38893-0050
Tributyltin trifluoromethanesulfonate 96%
Tributyltin trifluoromethanesulfonate
Formula: C₁₃H₂₇F₃O₃SSn
MW: 439.121 g/mol
Boiling Pt: 56…60 °C (25 mmHg)
Melting Pt: 31 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00043324
CAS Number: 68725-14-4
UN: 2923
ADR: 8,II

Order Now

Specification Test Results

Appearance White to light beige Crystals and/or chunks
Infrared spectrum Conforms
Titration with NaOH ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR