Isoindoline hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

AC43585-0050 AC43585-0250
TS43585-0050EA 161 USD
TS43585-0050 TS43585-0250
Isoindoline hydrochloride 97%
Isoindoline hydrochloride
Formula: C₈H₁₀ClN
Melting Pt: 255…256 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD03502377
CAS Number: 32372-82-0

Order Now

Specification Test Results

Appearance White to off-white to gray Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Elemental Analysis 8.64 to 9.36 % N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR