Silver lactate 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC23333-0050
AC23333-0100
AC23333-0500
TS23333-0050EA
75
USD
TS23333-0050
TS23333-0100
TS23333-0500
Silver lactate 96%
Silver lactate
Formula:
C₃H₅AgO₃ Melting Pt: 120…122 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043279 CAS Number: 15768-18-0 |
Specification Test Results
Appearance | Gray-brown Powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥95 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...