Calcium iodate, pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC31841-0025
AC31841-1000
AC31841-5000
TS31841-0025EA
430
USD
TS31841-0025
TS31841-1000
TS31841-5000
Calcium iodate, pure
Calcium iodate
Formula:
CaI₂O₆ Melting Pt: 540 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00036265 CAS Number: 7789-80-2 UN: 1479 ADR: 5.1,II |
Specification Test Results
Appearance | White to light beige Crystalline powder |
Titration Iodometric | ≥96 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...