3-Isocyanatopropyltriethoxysilane 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: ICPTES, Triethoxy(3-isocyanatopropyl)silane, (3-Isocyanatopropyl)triethoxysilane, Isocyanic acid 3-(Triethoxysilyl)propyl ester, 3-(Triethoxysilyl)propyl Isocyanate

AC39190-0250 AC39190-1000
TS39190-0250EA 53 USD
TS39190-0250 TS39190-1000
3-Isocyanatopropyltriethoxysilane 95%
3-Isocyanatopropyltriethoxysilane
Formula: C₁₀H₂₁NO₄Si
MW: 247.366 g/mol
Boiling Pt: 130 °C (20 mmHg)
Density: 0.99 g/cm³
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00051459
CAS Number: 24801-88-5
UN: 2927
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4240 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR