Sodium-tert-pentyloxide 95%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium tert-pentoxide, Sodium tert-pentoxide, 95%, tert-Amyl alcohol sodium salt

AC32755-1000 AC32755-5000
TS32755-1000EA 73 USD
TS32755-1000 TS32755-5000
Sodium-tert-pentyloxide 95%, pure
Sodium-tert-pentyloxide
Formula: C₅H₁₁NaO
MW: 110.132 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064245
CAS Number: 14593-46-5
UN: 3206
ADR: 4.2,II

Order Now

Specification Test Results

Appearance Cream to tan Powder
Titration with HClO4 ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR