Aluminum phosphate, pure
Supplier: Thermo Scientific Chemicals
AC34444-0025
AC34444-1000
AC34444-5000
TS34444-0025EA
432
USD
TS34444-0025
TS34444-1000
TS34444-5000
Aluminum phosphate, pure
Aluminum phosphate
Formula:
AlPO₄ Melting Pt: >1500 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003430 CAS Number: 7784-30-7 |
Specification Test Results
Appearance | White Powder |
Titration | ≥80.0 % (AlPO4) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...