tert-Butoxybis(dimethylamino)methane

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bredereck's reagent, 1-tert-Butoxy-N,N,N',N'-tetramethylmethylenenediamine, [(tert-butoxy)(dimethylamino)methyl]dimethylamine, tert-Butoxy bis(dimethylamino)methane

AC37812-0050 AC37812-0250
TS37812-0050EA 124 USD
TS37812-0050 TS37812-0250
tert-Butoxybis(dimethylamino)methane
tert-Butoxybis(dimethylamino)methane
Formula: C₉H₂₂N₂O
MW: 174.288 g/mol
Boiling Pt: 50…55 °C (15 mmHg)
Density: 0.844 g/cm³
Flash Pt: 41 °C
Storage Temperature: Freezer
MDL Number: MFCD00042858
CAS Number: 5815-08-7
UN: 1993
ADR: 3 III

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
GC 90 % (sum of 3 constituents) varying proportionally Tris(dimethylamino)methane, N,N-dimethylformamide-N,N-di-tert.-butylacetal, tert.-Butoxybis(dimethylamino)methane


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR