1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate 97%

Supplier: Thermo Scientific Chemicals
Warning

AC37794-0010 AC37794-2500
TS37794-0010EA 315 USD
TS37794-0010 TS37794-2500
1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate 97%
1,5-Cyclooctadienebis(methyldiphenylphosphine)iridium(I) hexafluorophosphate
Formula: C₃₄H₃₈F₆IrP₃
MW: 845.79 g/mol
Melting Pt: 224 °C
Storage Temperature: Ambient
MDL Number: MFCD00064800
CAS Number: 38465-86-0

Order Now

Specification Test Results

Appearance Red Crystalline powder
Infrared spectrum Conforms
Assay ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR