(2-Bromoethoxy)-tert-butyldimethylsilane 98+% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-Bromoethoxy)(tert-butyl)dimethylsilane

AC39983-0010 AC39983-0100
TS39983-0010EA 36 USD
TS39983-0010 TS39983-0100
(2-Bromoethoxy)-tert-butyldimethylsilane 98+% stabilized
(2-Bromoethoxy)-tert-butyldimethylsilane
Formula: C₈H₁₉BrOSi
MW: 239.228 g/mol
Boiling Pt: 70…75 °C (2.5 mmHg)
Density: 1.115 g/cm³
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00209550
CAS Number: 86864-60-0

Order Now

Specification Test Results

Appearance Colorless Liquid
Infrared spectrum Conforms
GC ≥98.0 %
Stabilizer ≤5 % Na2CO3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR