Tetra-tert-butyl orthotitanate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Titanium(IV) tert-butoxide, Titanic acid tetra-tert-butyl ester, Titanium(IV) t-butoxide, Titanium tetra-tert-butylate, Orthotitanic acid tetra-tert-butyl ester, tert-Butyl titanate(IV)

AC34400-0050 AC34400-0250
TS34400-0050EA 87 USD
TS34400-0050 TS34400-0250
Tetra-tert-butyl orthotitanate 98%
Tetra-tert-butyl orthotitanate
Formula: C₁₆H₃₆O₄Ti
MW: 340.32 g/mol
Boiling Pt: 160…165 °C (15 mmHg)
Density: 0.8893 g/cm³
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00040554
CAS Number: 3087-39-6
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Assay ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR