Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0) 96%

Supplier: Thermo Scientific Chemicals
Warning

AC43915-0010 AC43915-0050
TS43915-0010EA 80 USD
TS43915-0010 TS43915-0050
Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0) 96%
Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0)
Formula: C₄₂H₄₄O₁₀Pd
Melting Pt: 170…178 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07369799
CAS Number: 811862-77-8

Order Now

Specification Test Results

Appearance Red-brown Powder
Infrared spectrum Conforms
NMR ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR