4-(2-Aminoethyl)-2-methoxyphenol hydrochloride 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Methoxytyramine·HCl, 3-O-Methyldopamine hydrochloride

AC17285-0010 AC17285-1000
TS17285-0010EA 326 USD
TS17285-0010 TS17285-1000
4-(2-Aminoethyl)-2-methoxyphenol hydrochloride 99+%
4-(2-Aminoethyl)-2-methoxyphenol hydrochloride
Formula: C₉H₁₄ClNO₂
MW: 203.666 g/mol
Melting Pt: 213…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00012902
CAS Number: 1477-68-5

Order Now

Specification Test Results

Appearance Light beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR