Zirconium(IV) Propoxide ∼ 70% in propanol, AcroSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC42793-1000
AC42793-8000
TS42793-1000EA
41
USD
TS42793-1000
TS42793-8000
Zirconium(IV) Propoxide ∼ 70% in propanol, AcroSeal™
Zirconium(IV) Propoxide
Formula:
C₁₂H₂₈O₄Zr Density: 1.044 g/cm³ Flash Pt: 23 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015307 CAS Number: 23519-77-9 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Colorless to yellow to amber Solution |
Assay | 27.0 to 29.0 % (ZrO2) |
Melting point | 73 to 80 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...