Zinc(II) dibenzyldithiocarbamate 95%

Supplier: Thermo Scientific Chemicals
Warning

AC40644-0250 AC40644-1000
TS40644-0250EA 279 USD
TS40644-0250 TS40644-1000
Zinc(II) dibenzyldithiocarbamate 95%
Zinc(II) dibenzyldithiocarbamate
Formula: C₁₅H₁₅NS₂
Melting Pt: 183…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00014470
CAS Number: 14726-36-4
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 183 to 188 °C
Assay ≥94 % (ex Zn)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR