Vinylmagnesium bromide 1 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC44597-1000 AC44597-8000
TS44597-1000EA 125 USD
TS44597-1000 TS44597-8000
Vinylmagnesium bromide 1 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal™
Vinylmagnesium bromide
Formula: C₂H₃BrMg
MW: 131.26 g/mol
Density: 0.965 g/cm³
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00000042
CAS Number: 1826-67-1
UN: 3399
ADR: 4.3 I

Order Now

Specification Test Results

Appearance Yellow to red to brown Solution (May develop some turbidity or precipitate)
Titration Active base 0.80 to 1.10 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR