Trimethyl(vinyloxy)silane 97%

Supplier: Thermo Scientific Chemicals
Danger

AC33870-0010 AC33870-0050
TS33870-0010EA 34 USD
TS33870-0010 TS33870-0050
Trimethyl(vinyloxy)silane 97%
Trimethyl(vinyloxy)silane
Formula: C₅H₁₂OSi
MW: 116.24 g/mol
Boiling Pt: 74…75 °C (760 mmHg)
Density: 0.77 g/cm³
Flash Pt: –10 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00054764
CAS Number: 6213-94-1
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3885 to 1.3905 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR