Tetramethylammonium triacetoxyborohydride 90+%

Supplier: Thermo Scientific Chemicals
Danger

AC39285-0100 AC39285-0500
TS39285-0100EA 151 USD
TS39285-0100 TS39285-0500
Tetramethylammonium triacetoxyborohydride 90+%
Tetramethylammonium triacetoxyborohydride
Formula: C₁₀H₂₂BNO₆
Melting Pt: 95…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00012196
CAS Number: 109704-53-2
UN: 2813
ADR: 4.3,I

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Titration with HClO4 ≥90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR