tert-Butylmagnesium chloride 1.7 M in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC33167-1000 AC33167-8000
TS33167-1000EA 70 USD
TS33167-1000 TS33167-8000
tert-Butylmagnesium chloride 1.7 M in THF, AcroSeal™
tert-Butylmagnesium chloride
Formula: C₄H₉ClMg
MW: 116.87 g/mol
Density: 0.931 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00000465
CAS Number: 677-22-5
UN: 3399
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Dark brown Solution
Titration Active base 19 to 21 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR