tert-Butyldimethylsilane 95%

Supplier: Thermo Scientific Chemicals
Danger

AC44571-0050 AC44571-0250
TS44571-0050EA 84 USD
TS44571-0050 TS44571-0250
tert-Butyldimethylsilane 95%
tert-Butyldimethylsilane
Formula: C₆H₁₆Si
MW: 116.28 g/mol
Boiling Pt: 81…83 °C
Density: 0.701 g/cm³
Flash Pt: –11 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010754
CAS Number: 29681-57-0
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR