p-Tolylmagnesium bromide ∼0.5 M in diethyl ether, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC42676-1000 AC42676-8000
TS42676-1000EA 59 USD
TS42676-1000 TS42676-8000
p-Tolylmagnesium bromide ∼0.5 M in diethyl ether, AcroSeal™
p-Tolylmagnesium bromide
Formula: C₇H₇BrMg
Density: 0.866 g/cm³
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00000040
CAS Number: 4294-57-9
UN: 2924
ADR: 3,I

Order Now

Specification Test Results

Appearance Clear to hazy yellow to amber Solution (May develop some turbidity or precipitate)
Titration Grignard 0.40 to 0.60 M
Rhodium (Rh) ≥34.7 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR