Potassium trifluoro(trifluoromethyl)borate
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC46473-0010
464730050
TS46473-0010EA
106
USD
TS46473-0010
TS464730050
Potassium trifluoro(trifluoromethyl)borate
Potassium trifluoro(trifluoromethyl)borate
Formula:
CHBF₆ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD09038544 CAS Number: 42298-15-7 |
Specification Test Results
Appearance | White to yellow to brown Crystalline powder or solid |
Infrared spectrum | Conforms |
NMR (19F-NMR) | ≥96 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...