n-Octylmagnesium bromide 2 M in diethyl ether, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC42781-1000 AC42781-8000
TS42781-1000EA 79 USD
TS42781-1000 TS42781-8000
n-Octylmagnesium bromide 2 M in diethyl ether, AcroSeal™
n-Octylmagnesium bromide
Formula: C₈H₁₇BrMg
MW: 217.43 g/mol
Boiling Pt: 35 °C (760 mmHg)
Density: 0.961 g/cm³
Flash Pt: –35 °C
Storage Temperature: Ambient
MDL Number: MFCD00191566
CAS Number: 17049-49-9
UN: 2924
ADR: 3 I

Order Now

Specification Test Results

Appearance Yellow to brown Solution
Titration Grignard 1.9 to 2.2 M
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR