N-Carbobenzoxy-1,3-diaminopropane hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Benzyloxycarbonyl-1,3-propanediamine hydrochloride, N-Z-1,3-Propanediamine hydrochloride, N-Benzyloxycarbonyl-1,3-diaminopropane hydrochloride, N-Cbz-1,3-diaminopropane Hydrochloride, Benzyl N-(3-Aminopropyl)carbamate Hydrochloride, N-(3-Aminopropyl)carbamic Acid Benzyl Ester Hydrochloride, Benzyl (3-aminopropyl)carbamate hydrochloride

AC42707-0010 AC42707-0050
TS42707-0010EA 73 USD
TS42707-0010 TS42707-0050
N-Carbobenzoxy-1,3-diaminopropane hydrochloride 98%
N-Carbobenzoxy-1,3-diaminopropane hydrochloride
Formula: C₁₁H₁₆N₂O₂
MW: 244.72 g/mol
Melting Pt: 185…189 °C
Storage Temperature: Ambient
MDL Number: MFCD00270153
CAS Number: 17400-34-9

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 185 to 189 °C
NMR consistent with structure
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR