Benzyl N-(2-aminoethyl)carbamate hydrochloride 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC43981-0010
AC43981-0050
TS43981-0010EA
58
USD
TS43981-0010
TS43981-0050
Benzyl N-(2-aminoethyl)carbamate hydrochloride 95%
Benzyl N-(2-aminoethyl)carbamate hydrochloride
Formula:
C₁₀H₁₄N₂O₂ Melting Pt: 170…173 °C Storage Temperature: Ambient |
MDL Number:
MFCD00270150 CAS Number: 18807-71-1 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
HPLC | ≥94.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...