N,O-Bis(trimethylsilyl)hydroxylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

AC21598-0010 AC21598-0050 AC21598-0250
TS21598-0010EA 41 USD
TS21598-0010 TS21598-0050 TS21598-0250
N,O-Bis(trimethylsilyl)hydroxylamine 97%
N,O-Bis(trimethylsilyl)hydroxylamine
Formula: C₆H₁₉NOSi₂
Boiling Pt: 78…80 °C (100 mmHg)
Density: 0.83 g/cm³
Flash Pt: 28 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008266
CAS Number: 22737-37-7
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR