Hydridotetrakis(triphenylphosphine)rhodium(I)

Supplier: Thermo Scientific Chemicals

Synonyms: Tetrakis(triphenylphosphine)rhodium(I) Hydride

AC37793-0010 AC37793-2500
TS37793-0010EA 212 USD
TS37793-0010 TS37793-2500
Hydridotetrakis(triphenylphosphine)rhodium(I)
Hydridotetrakis(triphenylphosphine)rhodium(I)
Formula: C₇₂H₆₁P₄Rh
MW: 1153.08 g/mol
Melting Pt: 140…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00015867
CAS Number: 18284-36-1

Order Now

Specification Test Results

Appearance Ochre Crystalline powder
Infrared spectrum Conforms
Rhodium (Rh) ≥8.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR