Hexylmagnesium bromide 0.8 M in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC42776-1000 AC42776-8000
TS42776-1000EA 69 USD
TS42776-1000 TS42776-8000
Hexylmagnesium bromide 0.8 M in THF, AcroSeal™
Hexylmagnesium bromide
Formula: C₆H₁₃BrMg
MW: 189.38 g/mol
Density: 0.98 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000046
CAS Number: 3761-92-0
UN: 2924
ADR: 3 II

Order Now

Specification Test Results

Appearance Brown Solution
Titration Grignard 0.6 to 1 M
Melting point 62 to 65 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR