Ethylmagnesium chloride 25% (w/w) 2.7 M in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC25257-1000 AC25257-8000
TS25257-1000EA 53 USD
TS25257-1000 TS25257-8000
Ethylmagnesium chloride 25% (w/w) 2.7 M in THF, AcroSeal™
Ethylmagnesium chloride
Formula: C₂H₅ClMg
MW: 88.82 g/mol
Density: 1 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000472
CAS Number: 2386-64-3
UN: 3399
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Dark brown Solution
Titration with HCl 24 to 26 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR