Ethylenedioxybis(trimethylsilane) 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2-Bis(trimethylsilyloxy)ethane, Ethylene Glycol Bis(trimethylsilyl Ether), 1,2-Bis(trimethylsiloxy)ethane

AC20958-0100 AC20958-0500
TS20958-0100EA 51 USD
TS20958-0100 TS20958-0500
Ethylenedioxybis(trimethylsilane) 98%
Ethylenedioxybis(trimethylsilane)
Formula: C₈H₂₂O₂Si₂
MW: 206.43 g/mol
Boiling Pt: 165…166 °C (760 mmHg)
Density: 0.84 g/cm³
Flash Pt: 46 °C
Storage Temperature: Ambient
MDL Number: MFCD00009640
CAS Number: 7381-30-8
UN: 1993
ADR: 3 III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4020 to 1.4040 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR