Dichlorocyclohexylmethylsilane 97+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cyclohexylmethyldichlorosilane

AC43222-0100 AC43222-0500
TS43222-0100EA 85 USD
TS43222-0100 TS43222-0500
Dichlorocyclohexylmethylsilane 97+%
Dichlorocyclohexylmethylsilane
Formula: C₇H₁₄Cl₂Si
MW: 197.17 g/mol
Boiling Pt: 202 °C
Density: 1.095 g/cm³
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00046333
CAS Number: 5578-42-7
UN: 2987
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4710 to 1.4730 (20°C, 589 nm)
GC ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR