Dichloro(p-cymene)ruthenium(II) dimer 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Di(μ-chloro)dichlorobis(p-cymene)diruthenium(II)

AC36329-0010 AC36329-0050
TS36329-0010EA 69 USD
TS36329-0010 TS36329-0050
Dichloro(p-cymene)ruthenium(II) dimer 98%
Dichloro(p-cymene)ruthenium(II) dimer
Formula: C₂₀H₂₈Cl₄Ru₂
Melting Pt: 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064793
CAS Number: 52462-29-0

Order Now

Specification Test Results

Appearance Orange-red to red-brown Powder or crystals
Infrared spectrum Conforms
Elemental Analysis ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR