Dichloro(1,5-cyclooctadiene)platinum(II) 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,5-Cyclooctadieneplatinum(II) Dichloride
AC36361-0010
AC36361-2500
TS36361-0010EA
288
USD
TS36361-0010
TS36361-2500
Dichloro(1,5-cyclooctadiene)platinum(II) 98%
Dichloro(1,5-cyclooctadiene)platinum(II)
Formula:
C₈H₁₂Cl₂Pt MW: 374.17 g/mol Melting Pt: 285 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012413 CAS Number: 12080-32-9 |
Specification Test Results
Appearance | White to light yellow Crystalline powder or crystals |
Infrared spectrum | Conforms |
Assay | ≥97.5 % (ex Pt) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...