Cyclopentylmagnesium bromide 2.0 M in diethyl ether, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC42745-1000 AC42745-8000
TS42745-1000EA 117 USD
TS42745-1000 TS42745-8000
Cyclopentylmagnesium bromide 2.0 M in diethyl ether, AcroSeal™
Cyclopentylmagnesium bromide
Formula: C₅H₉BrMg
MW: 173.34 g/mol
Boiling Pt: 35 °C (760 mmHg)
Density: 0.996 g/cm³
Flash Pt: –38 °C
Storage Temperature: Ambient
MDL Number: MFCD00209569
CAS Number: 33240-34-5
UN: 3399
ADR: 4.3 I

Order Now

Specification Test Results

Appearance Gray Solution
Titration Grignard 1.90 to 2.20 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR