Copper(I) trifluoromethanesulfonate benzene complex 90%
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC37109-0010
AC37109-0050
TS37109-0010EA
64
USD
TS37109-0010
TS37109-0050
Copper(I) trifluoromethanesulfonate benzene complex 90%
Copper(I) trifluoromethanesulfonate benzene complex
Formula:
[CuOSO₂CF₃]₂.C₆H₆ Melting Pt: 160 °C (dec.) Flash Pt: 7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043300 CAS Number: 42152-46-5 UN: 1325 ADR: 4.1,III |
Specification Test Results
Appearance | Gray-green Powder |
Infrared spectrum | Conforms |
Titration with KMnO4 | ≥88 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...