Chlorobis(ethylene)rhodium(I) dimer

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bis(ethylene)rhodium(I) chloride dimer, [Rh(C₂H₄)₂Cl]₂, µ-Dichlorotetraethylene dirhodium(I), Dichlorotetra(ethylene)dirhodium(I), Dichlorotetrakis(ethylene)dirhodium(I)

AC19759-0010 AC19759-1000
TS19759-0010EA 727 USD
TS19759-0010 TS19759-1000
Chlorobis(ethylene)rhodium(I) dimer
Chlorobis(ethylene)rhodium(I) dimer
Formula: C₈H₁₆Cl₂Rh₂
MW: 388.93 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00013206
CAS Number: 12081-16-2

Order Now

Specification Test Results

Appearance Orange to rust brown to black Crystalline powder and/or chunks
Infrared spectrum Conforms
Rhodium (Rh) ≥52.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR