Chlorobis(cyclooctene)rhodium(I) dimer 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC36937-1000
AC36937-5000
TS36937-1000EA
70
USD
TS36937-1000
TS36937-5000
Chlorobis(cyclooctene)rhodium(I) dimer 98%
Chlorobis(cyclooctene)rhodium(I) dimer
Formula:
C₃₂H₅₆Cl₂Rh₂ Melting Pt: 187…191 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00013287 CAS Number: 12279-09-3 |
Specification Test Results
Appearance | Orange-brown Powder |
Infrared spectrum | Conforms |
Rhodium (Rh) | 28.0 to 29.4 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...