Carboxymethoxylamine hemihydrochloride 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Aminooxyacetic acid hemihydrochloride, O-(Carboxymethyl)hydroxylamine Hemihydrochloride, O-(Carboxymethyl)hydroxylamine hemihydrochloride, (Aminooxy)acetic Acid Hemihydrochloride

AC10843-0025 AC10843-0100 AC10843-0500
TS10843-0025EA 110 USD
TS10843-0025 TS10843-0100 TS10843-0500
Carboxymethoxylamine hemihydrochloride 98%
Carboxymethoxylamine hemihydrochloride
Formula: C₄H₁₁ClN₂O₆
Melting Pt: 156 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012955
CAS Number: 2921-14-4

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR