Bis(triphenylphosphine)palladium(II) diacetate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Bis(triphenylphosphine)palladium(II) acetate, Bis(acetato)bis(triphenylphosphine)palladium(II), Diacetatobis(triphenylphosphine)palladium(II)

AC20927-0010 AC20927-0050
TS20927-0010EA 102 USD
TS20927-0010 TS20927-0050
Bis(triphenylphosphine)palladium(II) diacetate 99%
Bis(triphenylphosphine)palladium(II) diacetate
Formula: C₄₀H₃₆O₄P₂Pd
MW: 749.09 g/mol
Melting Pt: 136 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010013
CAS Number: 14588-08-0

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
Assay ≥14 % (Pd)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR