Bis(tri-o-tolylphosphine)palladium(II) dichloride 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
trans-Dichlorobis(tri-o-tolylphosphine)palladium(II), Bis[tri(o-tolyl)phosphine]palladium(II) chloride, Dichlorobis(tri-o-tolylphosphine)palladium(II), Dichlorobis(tri-o-tolylphsophine)palladium
AC37796-0010
AC37796-0050
TS37796-0010EA
57
USD
TS37796-0010
TS37796-0050
Bis(tri-o-tolylphosphine)palladium(II) dichloride 95%
Bis(tri-o-tolylphosphine)palladium(II) dichloride
Formula:
C₄₂H₄₂Cl₂P₂Pd MW: 786.07 g/mol Melting Pt: 280 °C Storage Temperature: Ambient |
MDL Number:
MFCD00274659 CAS Number: 40691-33-6 |
Specification Test Results
Appearance | Yellow to orange Crystalline powder |
Infrared spectrum | Conforms |
Elemental Analysis | ≥94 % (ex C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...