Bis(trimethylsilyl)malonate 98%

Supplier: Thermo Scientific Chemicals
Warning

AC31330-0100 AC31330-0500
TS31330-0100EA 85 USD
TS31330-0100 TS31330-0500
Bis(trimethylsilyl)malonate 98%
Bis(trimethylsilyl)malonate
Formula: C₉H₂₀O₄Si₂
Boiling Pt: 63…66 °C (1 mmHg)
Density: 0.972 g/cm³
Flash Pt: 23 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009631
CAS Number: 18457-04-0
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to straw yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4150 to 1.4170 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR