Bis(tricyclohexylphosphine)palladium(II) dichloride 98%

Supplier: Thermo Scientific Chemicals
Warning

AC39589-0010 AC39589-2500
TS39589-0010EA 141 USD
TS39589-0010 TS39589-2500
Bis(tricyclohexylphosphine)palladium(II) dichloride 98%
Bis(tricyclohexylphosphine)palladium(II) dichloride
Formula: C₃₆H₆₆Cl₂P₂Pd
Melting Pt: 270 °C
Storage Temperature: Ambient
MDL Number: MFCD00191830
CAS Number: 29934-17-6

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
Elemental Analysis ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR