Benzylbis(triphenylphosphine)palladium(II) chloride

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (SP-4-3)-Benzylbis(triphenylphosphine)palladium(II) chloride, trans-Benzyl(chloro)bis(triphenylphosphine)palladium(II), Benzyl(chloro)bis(triphenylphosphine)palladium(II)

AC36971-1000 AC36971-5000
TS36971-1000EA 67 USD
TS36971-1000 TS36971-5000
Benzylbis(triphenylphosphine)palladium(II) chloride
Benzylbis(triphenylphosphine)palladium(II) chloride
Formula: C₄₃H₃₇ClP₂Pd
MW: 757.59 g/mol
Melting Pt: 166…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00010395
CAS Number: 22784-59-4

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Palladium (Pd) ≥13.8 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR